missing translation for 'onlineSavingsMsg'
Learn More

FKBP38 Rabbit anti-Human, Mouse, Rat, Clone: 2J7P6, Novus Biologicals™

Product Code. 18332247 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332247 100 μg 100µL
18357972 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332247 Supplier Novus Biologicals Supplier No. NBP316684100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FKBP38 Monoclonal antibody specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP38
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 2J7P6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FKBP38 (Q14318).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cellular Markers, Core ESC Like Genes, Glia Markers, Growth and Development, Mitotic Regulators, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 23770
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.