missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP25 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | FKBP25 |
|---|---|
| Dilution | Western Blot 1:500, Simple Western 1:25, ELISA 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18361874
|
Novus Biologicals
NBP3-18658-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18348174
|
Novus Biologicals
NBP3-18658-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP25 Polyclonal antibody specifically detects FKBP25 in Mouse samples. It is validated for Western Blot, Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| FKBP25 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| Whole antiserum | |
| EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
| Polyclonal antibody raised in rabbit against mouse FKBP25 (FK506 Binding Protein 3), using a KLHconjugated synthetic peptide containing an amino acid sequence from the central part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:500, Simple Western 1:25, ELISA 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| Q00688 | |
| 2287 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title