missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00022-100ul
This item is not returnable.
View return policy
Description
FKBP25 Polyclonal antibody specifically detects FKBP25 in Human samples. It is validated for ELISA
Specifications
| FKBP25 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
| Produced in rabbits immunized with purified, recombinant Human FKBP25 (Uniprot#: Q00688; Ala2-Asp224) | |
| 100 μg | |
| Stem Cell Markers | |
| 2287 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction