missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP25 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-41071H
This item is not returnable.
View return policy
Description
FKBP25 Polyclonal antibody specifically detects FKBP25 in Human samples. It is validated for Western Blot
Specifications
| FKBP25 | |
| Polyclonal | |
| Western Blot | |
| EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
| The immunogen for this product maps to a region between residue 50 and 100 of human FK506 binding protein 3 using the numbering given in entry NP_002004.1 (GeneID 2287). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 2287 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction