missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561958
This item is not returnable.
View return policy
Description
Immunogen sequence: RKLQIGVKKR VDHCPIKSRK GDVLHMHYT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
FKBP2 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is thought to function as an ER chaperone and may also act as a component of membrane cytoskeletal scaffolds.
Specifications
| FKBP2 | |
| Polyclonal | |
| Unconjugated | |
| Fkbp2 | |
| 13 kDa FK506-binding protein; 13 kDa FKBP; 13kDa; FK506 binding protein 2; FK506 binding protein 2, 13kDa; FK506-binding protein 2; FK506-binding protein 2 (13kD); FKBP prolyl isomerase 2; FKBP13; FKBP-13; FKBP2; FKBP-2; Immunophilin FKBP13; mFKBP13; mFKBP2; Peptidyl-prolyl cis-trans isomerase FKBP2; PPIase; PPIase FKBP2; proline isomerase; rapamycin-binding protein; RGD1560660; Rotamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26885 | |
| Fkbp2 | |
| Recombinant protein corresponding to Human FKBP2. Recombinant protein control fragment (Product #RP-103050). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction