missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FKBP1A |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856553
|
Proteintech
10273-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846553
|
Proteintech
10273-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunophilins are a family of soluble receptors capable of binding to one of two major immunosuppressant agents; cyclosporin A (CsA) or FK506. Proteins that bind FK506 are termed FK506 Binding Proteins (FKBPs) and those that bind cyclosporin A are called cyclophilins (CyP). Immunophilins function as cis-trans peptidyl-prolyl isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. Thus, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline-containing proteins. FKBP12:FK506 complexes inhibit calcineurin, a calcium/calmodulin-dependent serine/threonine phosphatase which blocks T-cell activation by preventing lymphokine gene transcription. FKBP12 also plays a role in intracellular calcium regulation by associating with three types of calcium release channel complexes, cardiac and skeletal ryanodine receptors and the inositol 1,4,5-trisphosphate receptor. In interactions with members of the TGF beta family type I receptors, FKBP12 has been shown to exert an inhibitory effect on the signaling pathway.Specifications
| FKBP1A | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P26883, P62942 | |
| 14225, 2280 | |
| FKBP1A Fusion Protein Ag0406 | |
| Primary | |
| -20°C | |
| FKBP1A |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 12 kDa FK506 binding protein, 12 kDa FKBP, FK506 binding protein 1A, FKBP 12, FKBP 1A, FKBP1, FKBP12, FKBP12C, FKBP1A, Immunophilin FKBP12, PKC12, PKCI2, PPIASE, PPIase FKBP1A, Rotamase | |
| FKBP1A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title