missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-82972-25ul
This item is not returnable.
View return policy
Description
FKBP15 Polyclonal specifically detects FKBP15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FKBP15 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| FK506 binding protein 15, 133kDa, FK506-binding protein 133kDa, FK506-binding protein 15, FKBP133, FKBP-133, FKBP-15,133 kDa FKBP, PPP1R76,133 kDa FK506-binding protein, regulatory subunit 76, WAFL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FKBP15 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KHSAGNSMLIPSMSVTMETSMIMSNIQRIIQENERLKQEILEKSNRIEEQNDKISELIERNQRYVEQSNLMMEKRNNSLQTATENTQARVLHAEQEKAKVTEELAAATAQVSHLQLKMTAHQ | |
| 25 μL | |
| Signal Transduction | |
| 23307 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction