Learn More
Invitrogen™ FKBP14 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100529
Product Code. 16627014
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total FKBP14.
PPIases accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FKBP14
Polyclonal
Unconjugated
Fkbp14
22 kDa FK506-binding protein; 22 kDa FKBP; BC029109; EDSKMH; FK506 binding protein 14; FK506 binding protein 14, 22 kDa; FK506-binding protein 14; Fkbp14; FKBP-14; FKBP-2; FKBP22; FKBP-22; IPBP12; peptidyl-prolyl cis-trans isomerase FKBP14; PPIase; PPIase FKBP14; rotamase; UNQ322/PRO381
Rabbit
Affinity chromatography
RUO
231997, 362366, 55033
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P59024, Q9NWM8
Fkbp14
A synthesized peptide derived from human FKBP14(Accession Q9NWM8), corresponding to amino acid residues G115-K165.
100 μL
Primary
Human, Mouse, Rat