missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP14 Mouse anti-Human, Clone: 1.80E+03, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59474-50UL
This item is not returnable.
View return policy
Description
FKBP14 Monoclonal specifically detects FKBP14 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FKBP14 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| 22 kDa FK506-binding protein, EC 5.2.1.8, FK506 binding protein 14 (22 kDa), FK506 binding protein 14, 22 kDa, FK506-binding protein 14, FKBP-14, FKBP-22, FKBP22PPIase FKBP14, FLJ20731, peptidyl-prolyl cis-trans isomerase FKBP14,22 kDa FKBP, rotamase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 55033 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1800 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| FKBP14 | |
| Recombinant human FKBP14 (20-211aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction