missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FKBP14 Monoclonal antibody specifically detects FKBP14 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Immunogen | Recombinant human FKBP14 (20-211aa) purified from E. coli |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?