missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP13/FKBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | FKBP13/FKBP2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658885
|
Novus Biologicals
NBP2-38604-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18136339
|
Novus Biologicals
NBP2-38604 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP13/FKBP2 Polyclonal specifically detects FKBP13/FKBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FKBP13/FKBP2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P26885 | |
| 2286 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RKLQIGVKKRVDHCPIKSRKGDVLHMHYT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 5.2.1.8,13 kDa FK506-binding protein, FK506 binding protein 2, 13kDa, FK506-binding protein 2, FK506-binding protein 2 (13kD), FKBP13,13 kDa FKBP, FKBP-13proline isomerase, FKBP-2, Immunophilin FKBP13, peptidyl-prolyl cis-trans isomerase FKBP2, PPIase, PPIase FKBP2, rapamycin-binding protein, Rotamase | |
| FKBP2 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title