missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP13/FKBP2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97373IR
This item is not returnable.
View return policy
Description
FKBP13/FKBP2 Polyclonal antibody specifically detects FKBP13/FKBP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FKBP13/FKBP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| EC 5.2.1.8,13 kDa FK506-binding protein, FK506 binding protein 2, 13kDa, FK506-binding protein 2, FK506-binding protein 2 (13kD), FKBP13,13 kDa FKBP, FKBP-13proline isomerase, FKBP-2, Immunophilin FKBP13, peptidyl-prolyl cis-trans isomerase FKBP2, PPIase, PPIase FKBP2, rapamycin-binding protein, Rotamase | |
| Produced in rabbits immunized with E. coli-derived Human FKBP13/FKBP2 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 2286 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction