missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FKBP12 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA514852

Product Code. 10282600

  • £479.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology.

FKBP12 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It interacts with several intracellular signal transduction proteins including type I TGF-beta receptor. It also interacts with multiple intracellular calcium release channels, and coordinates multi-protein complex formation of the tetrameric skeletal muscle ryanodine receptor. In mouse, deletion of this homologous gene causes congenital heart disorder known as noncompaction of left ventricular myocardium.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FKBP12
Polyclonal
Unconjugated
FKBP1A
12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; calstabin-1; EC 5.2.1.8; FK506 binding protein 1a; FK506 binding protein 1A, 12kDa; FK506 binding protein 2 (13 kDa); FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 1 (12kD); FK506-binding protein 12; FK506-binding protein 1a; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; Fkbp; FKBP1; FKBP12; FKBP-12; FKBP12-Exip3; Fkbp1a; FKBP-1A; Fkbp2; Immunophilin FKBP12; peptidyl-prolyl cis-trans isomerase FKBP1A; PKC12; PKCI2; PPIASE; PPIase FKBP1A; protein kinase C inhibitor 2; rotamase
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
2280
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P62942
FKBP1A
KLH conjugated synthetic peptide between 43-74 amino acids from the central region of human FKBP12.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.