missing translation for 'onlineSavingsMsg'
Learn More

FKBP12 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18399413 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399413 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399413 Supplier Novus Biologicals Supplier No. NBP299149R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FKBP12 Polyclonal specifically detects FKBP12 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FKBP12 (NP_463460; Met 1-Glu 108).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2280
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.