missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41272CL1
This item is not returnable.
View return policy
Description
FKBP12 Polyclonal antibody specifically detects FKBP12 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| FKBP12 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Antibody was raised against an 18 amino acid peptide near the center of human FKBP12. The immunogen is located within the first 50 amino acids of FKBP12. Amino Acid Squence: MLEDGKKFDSSRDRNKPF | |
| 0.1 mL | |
| Autophagy, Cancer, mTOR Pathway | |
| 2280 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction