missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | FKBP12 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686936
|
Novus Biologicals
NBP2-41272-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18185048
|
Novus Biologicals
NBP2-41272 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP12 Polyclonal specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| FKBP12 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| FKBP1A | |
| IgG | |
| Affinity Purified | |
| 12 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cancer, mTOR Pathway | |
| PBS with 0.02% Sodium Azide | |
| 2280 | |
| Antibody was raised against an 18 amino acid peptide near the center of human FKBP12. The immunogen is located within the first 50 amino acids of FKBP12. Amino Acid Squence: MLEDGKKFDSSRDRNKPF | |
| Primary | |
| FKBP12 antibody is human, mouse and rat reactive. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title