Learn More
Invitrogen™ FKBP10 Recombinant Rabbit Monoclonal Antibody (2M7X3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558163
Description
Immunogen sequence: FDSSYSRNHT YNTYIGQGYI IPGMDQGLQG ACMGERRRIT IPPHLAYGEN GTGDKIPGSA VLIFNVHVID FHNPADVVEI RTLSRPSETC NETTKLGDFV RYHYNCSLLD GTQLFTSHDY GAPQEATLGA NKVIEGLDTG LQGMCVGERR QLIVPPHLAH GESGARGVPG SAVLLFEVEL VSREDGLPTG YLFVWHKDPP ANLFEDMDLN KDGEVPPEEF STFIKAQVSE GKGRLMPGQD PEKTIGDMFQ NQDRN.
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.
Specifications
| FKBP10 | |
| Recombinant Monoclonal | |
| 1.35 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q61576 | |
| FKBP10 | |
| Recombinant protein (or fragment). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 2M7X3 | |
| Unconjugated | |
| FKBP10 | |
| 65 kDa FK506-binding protein; 65 kDa FKBP; AI325255; BRKS1; FK506 binding protein 10; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP10; FKBP-10; Fkbp1-rs; Fkbp6; FKBP65; FKBP-65; Fkbprp; Fkbp-rs; Fkbp-rs1; hFKBP65; Immunophilin FKBP65; OI11; OI6; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIASE; PPIase FKBP10; PSEC0056; Rotamase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14230 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.