missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106293
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FKBP65.
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.
Specifications
| FKBP10 | |
| Polyclonal | |
| Unconjugated | |
| FKBP10 | |
| 65 kDa FK506-binding protein; 65 kDa FKBP; AI325255; BRKS1; FK506 binding protein 10; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP10; FKBP-10; Fkbp1-rs; Fkbp6; FKBP65; FKBP-65; Fkbprp; Fkbp-rs; Fkbp-rs1; hFKBP65; Immunophilin FKBP65; OI11; OI6; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIASE; PPIase FKBP10; PSEC0056; Rotamase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14230, 360627, 60681 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61576, Q96AY3 | |
| FKBP10 | |
| A synthesized peptide derived from human FKBP10(Accession Q96AY3), corresponding to amino acid residues L218-V268. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction