missing translation for 'onlineSavingsMsg'
Learn More

FK506 Antibody (FK1), Novus Biologicals™

Product Code. 18053196 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053196 0.025 mg 25µg
18019945 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18053196 Supplier Novus Biologicals Supplier No. NB100625710.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FK506 Monoclonal specifically detects FK506 in Non-species specific samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FK506
Applications ELISA
Classification Monoclonal
Clone FK1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation TBS with 0.09% Sodium Azide
Host Species Mouse
Immunogen FK-506-BSA conjugate
Purification Method IgM purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62571 detects FK-506, a macrolide with similar but more potent immunosuppressant activity than cyclosporine. It is a 23-membered lactone produced by the bacteria Streptomyces tsukubaensis. FK-506 inhibits signal transduction and interleukin-2 transcription in CD4+ T helper lymphocytes, and is mainly used after allogenic organ transplantation to reduce organ rejection. Side effects of FK-506 treatment include nephrotoxicity, neurotoxicity, diabetes mellitus and hypertension.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.