missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FK 506
GREENER_CHOICE

Product Code. 18756286
Change view
Click to view available options
:
10mg
Product Code.
18756286 10mg
1 options
This item is not returnable. View return policy

Product Code. 18756286

Brand: Tocris Bioscience™ 3631/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent calcineurin inhibitor; immunosuppressant

FK 506 is a potent calcineurin (protein phosphatase 2B) inhibitor that requires FK 506-binding protein 12 (FKBP12) for activity (IC50 = 3 nM). Inhibits secretion of IL-1, IL-2 (IC50 = 1 nM), IL-3, IL-4, IL-6 (IC50 = 35 nM), GM-CSF, TNFα (IC50 = 10 nM), IFNγ and Myc from activated T cells in vitro. Exhibits potent immunosuppressive, neuroprotective and anticonvulsant activity in vivo. Identified as targeting human host cell proteins that interact with SARS-CoV-2.

Analog also available.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type FK 506
Purity >99%
Molecular Weight (g/mol) 804.02
Inhibitors FK 506

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.