missing translation for 'onlineSavingsMsg'
Learn More

FK 3311, MedChemExpress

Product Code. 16492207
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16492207

Brand: Medchem Express HY14445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FK 3311 (COX-2 Inhibitor V) is a selective inhibitor of COX-2 with antiinflammatory agent.

FK 3311 (COX-2 Inhibitor V) is a selective inhibitor of COX-2 with antiinflammatory agent.

Specifications

Chemical Name or Material FK 3311
CAS 116686-15-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H13F2NO4S
Quantity 50 mg
Synonym COX-2 Inhibitor V
Solubility Information DMSO : ≥ 100 mg/mL (292.97 mM)
SMILES FC1=CC=C(OC2=C(NS(=O)(C)=O)C=CC(C(C)=O)=C2)C(F)=C1
Molecular Weight (g/mol) 341.33
Formula Weight 341.33
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.