missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FJX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA563687
This item is not returnable.
View return policy
Description
Immunogen sequence: LALARVEARG AQWAQVQEEL RAAHWTEGSV VSLTRWLPNL TDVVVPAPWR SEDGRLRPLR DAGGELANLS QAELVDLVQW T Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
FJX1 Polyclonal antibody specifically detects FJX1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications
| FJX1 | |
| Polyclonal | |
| Unconjugated | |
| FJX1 | |
| FJX1; FLJ22416; FLJ25593; four jointed box 1; four jointed box 1 (Drosophila); four-jointed box kinase 1; four-jointed box protein 1; Four-jointed protein homolog; four-jointed protein x1; LOW QUALITY PROTEIN: four-jointed box protein 1; putative secreted ligand homologous to fjx1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24147 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VR8 | |
| FJX1 | |
| Recombinant protein corresponding to Human FJX1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction