missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FIZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540308
This item is not returnable.
View return policy
Description
Peptide sequence: HCGALYAALA ALEEHRRVSH GEGGGEEAAA AAREREPASG EPPSGSGRGK Sequence homology: Dog: 86%; Horse: 79%; Human: 100%; Pig: 83%.
FLJ14768 is homologous to mouse and bovine Fiz1 (flt-3 interacting zinc finger). It interacts with NRL (neural retina leucine zipper). Possible role for Fiz1 as a transcriptional repressor.
Specifications
| FIZ1 | |
| Polyclonal | |
| Unconjugated | |
| FIZ1 | |
| AI790204; FIZ1; FLT3 interacting zinc finger 1; Flt3 interacting zinc finger protein 1; FLT3-interacting zinc finger 1; flt3-interacting zinc finger protein 1; mFLJ00416; RGD1306359; Zinc finger protein 798; ZNF798 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 84922 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96SL8 | |
| FIZ1 | |
| Synthetic peptide directed towards the C-terminal of human FLJ14768. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction