Learn More
Microspec™ Fixed Volume Pipette
Description
- Fixed volume
- Single channel
- Non-sterile
- Grey
Specifications
Specifications
| Accuracy | 0.01 |
| Autoclavable | Not Autoclavable |
| Color | Gray |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Fixed Volume Pipette |
| Volume (Metric) | 5 μL |