Learn More
Erlab Fixed Shelf - Captair 714
Product Code. 15585194
Change view
Click to view available options
For Use With (Equipment):
Fuming Chamber
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15585194 Fuming Chamber Each
1 options
Product Code. 15585194 Supplier Erlab Supplier No. I0666000001
  • ONLINE EXCLUSIVE PRICE
    £452.00
    £443.92 / Pack of 1
    Save £8.08
    1% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
To store daily-use equipment.
  • Fixed Shelf for Benchcap for your ductless fume hood Captair 714
  • Intended Use: To store daily-use equipment
Specifications
For Use With (Equipment) Fuming Chamber
Product Type Shelf