Learn More
Thermo Scientific Fixed Price Preventative Maintenance for Fisherbrand Biosafety Cabinets
Product Code. 30503419
Change view
Click to view available options
For Use With (Equipment):
Biosafety Cabinets
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
30503419 Biosafety Cabinets Each
1 options
Product Code. 30503419 Supplier Thermo Scientific Supplier No. FPPMBSCFIS
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fixed-cost planned maintenance for Fisherbrand biosafety cabinets, including KI Discus test where required by national regulations.
  • In accordance with the European Standard for microbiological safety cabinets (CEN BS EN12469:2000, Table 5), the KI Discus test is identified as an optional test for operator/personal protection.
  • Applies to both commissioning and annual routine maintenance testing.
  • National regulations must be followed in all cases.
  • Some national regulations, such as those from the HSE (UK), recommend performing the KI Discus test at least annually.
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Biosafety Cabinets
    Product Type Preventive Maintenance