Learn More
Thermo Scientific™ Fixed Needle Gastight Autosampler Syringe
Product Code. 17874961 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
22 G
23 G
Volume (Metric):
1 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Volume (Metric) Unit Size
17874961 22 G 1 μL Each
16668853 23 G 1 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17874961 Supplier Thermo Scientific™ Supplier No. 365H3572
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Syringe for Thermo RSH autosampler
  • Engineered for dependable laboratory performance and consistent results
  • Manufactured to meet Thermo Scientific quality standards
  • Designed for compatibility with common laboratory instruments and workflows
  • Helps improve reproducibility and sample integrity
  • Suitable for research, analytical, and routine laboratory applications
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (Barrel)
Needle Gauge 22 G
Product Type Syringe
Volume (Metric) 1 μL
For Use With (Equipment) CTC PAL Liquid Autosampler
Connection Type Fixed, Tapered Needle
Graduated Yes
Sterility Non-sterile