Learn More
3M™ Fixed Length Breathing Tube SS-BT-23
Product Code. 17736973
Change view
Click to view available options
Breathing Tube; For Use With: 3M™ PAPR Systems
Product Code.
17736973 Breathing Tube; For Use With: 3M™ PAPR Systems
1 options
Product Code. 17736973 Supplier 3M™ Supplier No. 7100236823
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Fixed Length Breathing Tube SS-BT-23 is a fixed-length polyurethane breathing tube for use with approved 3M™ Powered Air Systems.
  • Fixed-length breathing tube made from polyurethane
  • Approved for use with 3M™ Powered Air Respirator PF-600E
  • Compatible with specific 3M™ Versaflo™ headtops when using the 3M™ Adapter Assembly QRS Thread ADP-03.
  • Protective breathing tube covers available to lengthen service life of breathing tubes