missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene Fixed High Intensity Multiplexing LEDs with Peak Band Passes
Shop All Syngene Products
Click to view available options
Color:
Blue
Green
Red, Green, Blue, Infrared, Far Red
Unit Size:
Each
Description
- HILED modules provide imaging performance for fluorescence applications.
- Choose from six options ranging from UV to IR light.
- Each module has a narrow band pass filter to minimise cross-talk between different wavelengths making it ideal for multiplexing with fluorescent gels or blots.
- Syngene GBox chemi systems are all compatible with these modules.
- The new HILED modules offer reduced exposure time, an extended LED life, and a 20 x 20 cm viewing area.
Spécification
Spécification
| Product Line | Syngene Gel Doc |
| For Use With (Application) | Imaging DNA Gels and Multiplexing |
| Color | Green |
| Product Type | HILEDS in Green |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu