missing translation for 'onlineSavingsMsg'
Learn More

Syngene Fixed High Intensity Multiplexing LEDs with Peak Band Passes

Product Code. 17872766 Shop All Syngene Products
Click to view available options
Color:
Blue
Green
Red, Green, Blue, Infrared, Far Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17872766

Brand: Syngene HILEDRGBIRFR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HILED modules provide imaging performance for fluorescence applications. There are six options that range from UV to IR light. Each module has a narrow band pass filter to minimise cross-talk ideal for multiplexing with fluorescent gels or blots.

  • HILED modules provide imaging performance for fluorescence applications.
  • Choose from six options ranging from UV to IR light.
  • Each module has a narrow band pass filter to minimise cross-talk between different wavelengths making it ideal for multiplexing with fluorescent gels or blots.
  • Syngene GBox chemi systems are all compatible with these modules.
  • The new HILED modules offer reduced exposure time, an extended LED life, and a 20 x 20 cm viewing area.

Specifications

Product Line Syngene Gel Doc
For Use With (Application) Imaging DNA Gels and Multiplexing
Color Red, Green, Blue, Infrared, Far Red
Product Type HILEDS in Red, Green, Blue, infra Red and Far Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.