Learn More
Syngene Fixed High Intensity Multiplexing LEDs
Product Code. 17944301 Shop All Syngene Products
Change view
Click to view available options
Product Type:
HILEDS
Product Code. Product Type
17944301 HILEDS
1 options
Product Code. 17944301 Supplier Syngene Supplier No. HILEDGRFR
Fixed high intensity multiplexing LEDs with peak band passes of red, green, blue, infra-red, and far-red.
Fixed high intensity multiplexing LEDs with peak band passes of red (623 nm), green (5205 nm), blue (465 nm), infra-red (740 nm), and far-red (638 nm).
Specifications
Product Type HILEDS