Learn More
VELP Scientifica FIWE Advance Automatic Fiber Analyzer
Product Code. 17851849 Shop All Velp Products
Change view
Click to view available options
Voltage:
115/230 V
Unit Size:
Each
Product Code. Voltage Unit Size
17851849 115/230 V Each
1 options
Product Code. 17851849 Supplier VELP Scientifica Supplier No. F30500500
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Fully automatic analyzer for crude and detergent fiber determination according to official reference methods requiring minimum operator time with unique user interface and cloud connectivity.
COMPLIANCE AND CONSISTENCY
  • The crucible method is adherent to official standards (ISO and AOAC) ensuring reliability and efficiency.
  • Crude Fiber determination and Detergent Fiber determination of NDF, ADF and ADL.
  • Fiber residues stay in crucibles during all steps of the analysis eliminating the risk of errors and ensuring consistent results.
LOAD THE SAMPLES AND GO
  • The FIWE Advance heats and doses the required reagents precisely to analyze up to 6 samples.
  • The VELP Dispensing Nozzle drives the reagents precisely into each column avoiding user contact with hot chemicals and fumes.
  • FIWE Advance automates digestion, filtration and washing steps without any operator intervention.
SAVING TIME AND MONEY
  • The operator time required is as little as 2 minutes.
  • The FIWE Advance treats each sample individually and takes care of each step of the analysis allowing for overnight work.
  • The FIWE Advance automatic processes free up valuable time enhancing lab productivity and saving money.
COMPLETE SAFETY
  • All the reagents required are contained in dedicated glass tanks and bottles located inside the instrument.
  • The FIWE Advance pre-heats, dispenses, and collects hot chemicals automatically so the risk of contact with the operator is eliminated.
SMART AND EASY TO USE
  • The 7 in. color touch screen offers simple and intuitive user interface designed to simplify operations.
  • Choose from preset methods or customize your own, mark as your favorites for quick access.
  • Optional balance connection and barcode scanner are available to simplify analysis preparation.
Specifications
Type Automatic Fiber Analyzer
Voltage 115/230 V