Learn More
Mettler Toledo™ LE621 Sensor for FiveGo™ Dissolved Oxygen Meters
Shop All Mettler Toledo Products
Click to view available options
:
LE621 sensor; IP67 waterproof
Description
- Robust and easy to use
- Plug/Connection: BNC/Cinch
Specifications
Specifications
| Connector Types | BNC/RCA (Cinch) IP67 |
| Description | LE DO Electrode LE621-IP67 |
| Type | LE621 Sensor, IP67 Waterproof |
| Includes | Integrated temperature sensor |
| Length (Metric) Cable | 1.00 m |