Learn More
Mettler Toledo™ LE703 Sensor for FiveGo™ Conductivity Meters
Product Code. 15553380 Shop All Mettler Toledo Products
Change view
Click to view available options
Length (Metric) Cable:
2 m
Unit Size:
Each
Product Code. Length (Metric) Cable unitSize
15553380 2 m Each
1 options
Product Code. 15553380 Supplier Mettler Toledo™ Supplier No. 30266974
Only null left
Add to basket
Add to basket
IP67 waterproof, robust conductivity probe for medium to high conductivities.
  • Range: 10μS/cm to 200mS/cm
  • Plug/Connection: LTW
TRUSTED_SUSTAINABILITY
Specifications
Connector Types LTW (IP67)
Description LE Cond Electrode LE703-IP67
Type LE703 Sensor, IP67 Waterproof
For Use With (Application) For medium to high conductivities
Length (Metric) Cable 2 m