missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIV Envelope Antibody (CE4-13B1 (13B1)), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66553G-0.1ML
This item is not returnable.
View return policy
Description
FIV Envelope Monoclonal specifically detects FIV Envelope in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FIV Envelope | |
| Monoclonal | |
| DyLight 488 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Purified gp95 envelope antigen. | |
| Primary | |
| Virus | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| CE4-13B1 (13B1) | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| 0.1 mL | |
| NB100-66553 is specific for FIV Envelope gp95 protein. This antibody is directed against an epitope within the variable V2 region and cross-reacts with Petaluma, 34TF10, Ppr, MCH5-5 and Glasgow 8 isolates. NB100-66553 does not neutralize FIV and does not recognize Feline Leukaemia Virus, Feline Herpes Virus Type 1, Feline Coronavirus and Feline Calicivirus. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction