Learn More
Cole-Parmer Fittings, Sanitary to Monobarb Adapter, Polypropylene
Product Code. p-200105082 Shop All Cole-Parmer Products
Click to view available options
Hose Barb:
1 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
Includes:
1-tri-clamp
3/4 in. Sanitary Clamp Size
Unit Size:
Each
Product Code. 30053803
Brand: Cole-Parmer 5011905
Only null left
Add to basket
Add to basket
High-quality nonmetallic sanitary fittings for critical fluid applications
Meet US and EU standards for use in pharmaceutical process applications. ASME-BPE, USP Class VI, and FDA compliant. Sterilize by autoclave, EtO, or gamma irradiation. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance ADCF;FDA;Non-Phthalate;RoHS;USP
Min. Operating Temperature (Metric) 0°C
Hose Barb 1/2 in.
Outlet Port Sanitary x Hose barb
Sterilization Method Autoclave, EtO, gamma irradiation
Includes 3/4 in. Sanitary Clamp Size
Max. Operating Temperature (Metric) 135°C