Learn More
Restek™ Fittings, Parker (Treated): Unions
Click to view available options
Type:
NPT Female Union
NPT Male Union
Reducing Union
Union
Material:
Silcosteel-CR Coated
Sulfinert-coated
Description:
1/16 in.
1/16 in. to 1/4 in.
1/16 in. to 1/8 in.
1/4 in.
1/4 in. to 1/4 in.
1/4 in. to 1/8 in.
1/8 in.
1/8 in. to 1/4 in.
1/8 in. to 1/8 in.
Description
- Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Specifications
| Type | Reducing Union |
| Description | 1/16 in. to 1/4 in. |
| Material | Sulfinert-coated |
| For Use With (Equipment) | Chromatography Connectors |