Learn More
Restek™ Fittings, Parker (Treated): End Reducer Tubes
Product Code. 13169492
Encompass_Preferred
Change view
Click to view available options
Connections:
1/4 in. to 1/16 in.
1/4 in. to 1/8 in.
1/8 in. to 1/16 in.
1/8 in. to 1/4 in.
Material:
Silcosteel-CR Coated
Sulfinert-coated
Unit Size:
Each
Product Code. Connections Material unitSize
13169492 1/4 in. to 1/16 in. Sulfinert-coated -
13159492 1/8 in. to 1/16 in. Sulfinert-coated -
13179492 1/8 in. to 1/4 in. Sulfinert-coated -
13189492 1/4 in. to 1/8 in. Sulfinert-coated -
13137292 1/8 in. to 1/16 in. Silcosteel-CR Coated Each
13147292 1/4 in. to 1/16 in. Silcosteel-CR Coated -
13167292 1/4 in. to 1/8 in. Silcosteel-CR Coated -
13157292 1/8 in. to 1/4 in. Silcosteel-CR Coated -
8 options
Product Code. 13169492
Brand: Restek™ 21545
This item requires a price update prior to being ordered.
Request a Price for this item here.
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Type Tube End Reducer
Connections 1/4 in. to 1/16 in.
Material Sulfinert-coated