Learn More
VICI Valco Instruments™ Fitting Eco-Cap
Brand:  VICI Valco Instruments™ JR-9000-0007
Code : 15
Additional Details : Weight : 1.00000kg
Product Code. 12702690
  • £89.78 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • A competitively priced alternative to sparge and deliver mobile phases
  • Includes tubing and filter
  • The Eco Cap consists of a three-hole ETFE insert (two for 1/8 in. and one for 4 mm tubing) with a polyethylene collar, compatible with all standard GL45 thread bottles.
  • The Eco Cap has no threading for fittings on the inside of the tubing ports – the tubing is simply inserted.
Specifications
Specifications
1 Female Luer Plug
Polyethylene