missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA185439
Code : LD
Additional Details : Weight : 1.24740kg
Description
Rabbit anti FITC antibody recognizes fluorescein and does not react with immunoglobulins from any species.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs μgtraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Specifications
FITC | |
Polyclonal | |
Unconjugated | |
Fluorescein | |
FITC Isomer 1. | |
1 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA, Immunohistochemistry (Frozen) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG |