missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Monoclonal Antibody (#8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514709
Code : LE
Additional Details : Weight : 0.01000kg
Description
Product MA514709 is a smaller package size of MIF2901 (formerly sold as a Seradyn product).
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs μgtraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Specifications
FITC | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
#8 | |
Unconjugated | |
Fluorescein | |
FITC-BSA | |
100 μg | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid | |
Chemical |