missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FITC/5-FAM/6-FAM Recombinant Rabbit Monoclonal Antibody (6R1B5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555270
This item is not returnable.
View return policy
Description
Immunogen Sequence: FITC conjugated to KLH.
FITC (Fluorescein Isothiocyanate), 5-FAM (5-Carboxyfluorescein), and 6-FAM (6-Carboxyfluorescein) are commonly used gene targets for fluorescent labeling in molecular biology research. These dyes belong to the fluorescein family and emit a green fluorescence when excited by specific wavelengths of light. FITC, 5-FAM, and 6-FAM are utilized in various techniques such as fluorescence in situ hybridization (FISH), flow cytometry, immunofluorescence, and DNA sequencing. By attaching these dyes to gene-specific probes or primers, researchers can visualize and detect gene expression levels, identify specific DNA or RNA sequences, and study cellular processes. The compatibility of FITC, 5-FAM, and 6-FAM with different imaging systems and their bright green fluorescence make them valuable tools in molecular biology, aiding in the understanding of gene function and cellular dynamics.
Specifications
| FITC/5-FAM/6-FAM | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Chemical compounds corresponding to FITC/5-FAM/6-FAM. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 6R1B5 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction