Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
£32.15 - £177.00
Specifications
Solution Type | Calibration Solution |
---|---|
Type | Conductivity Standard |
Certifications/Compliance | ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material |
Product Code | Brand | Description | Conductivity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Conductivity | Price | Quantity & Availability | ||||
11754236
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.000005 μ℧ | 200,000 μS |
£86.20
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11744236
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.000006 μ℧ | 150,000 μS |
£86.20
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11784226
|
Fisherbrand
|
NIST batch tested; Resistivity: 0.00001 mΩ | 100,000 μS |
£55.60
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11704246
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.00001 mΩ | 100,000 μS |
£86.20
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11774226
|
Fisherbrand
|
NIST batch tested; Resistivity: 0.0001 mΩ | 10,000 μS |
£54.65
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11794236
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.0001 mΩ | 10,000 μS |
£86.20
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11724246
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.00071 mΩ | 1413 μS |
£177.00
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11784236
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.001 mΩ | 1000 μS |
£87.70
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11754226
|
Fisherbrand
|
NIST batch tested; Resistivity: 0.01 mΩ | 100 μS |
£56.30
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11774236
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.01 mΩ | 100 μS |
£86.20
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11794226
|
Fisherbrand
|
NIST batch tested; Resistivity: 150000 μ℧ | 150,000 μS |
£32.15
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11704236
|
Fisherbrand
|
NIST batch tested; Resistivity: 200000 μ℧ | 200,000 μS |
£45.05
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11714246
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 0.5 mΩ | 5 μS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12399325
|
Fisherbrand
|
NIST Individual bottle tested; Resistivity: 1.0 mΩ | 1 μS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11714236
|
Fisherbrand
|
NIST batch tested; Resistivity: 0.00071 mΩ | 1413 μS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
- Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
- Also includes step-by step calibration instructions and temperature compensation chart
- Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
- ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Order Info
In 473mL (16 oz.) bottle
Specifications
Calibration Solution | |
Conductivity Standard | |
ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material |