missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

Brand:  Fisherbrand

Code : 46

Additional Details : Weight : 0.54000kg


 View more versions of this product

Product Code. 11774226

  • £54.65 / Each
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material

Order Info

473 mL (16 oz.) Bottle

Specifications

Specifications

NIST Batch Tested, Resistivity: 0.0001 mΩ
1 x 473 mL (16 oz.)
Clear
Conductivity Standard
ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Bottle
Calibration Solution
NIST Batch Tested
10,000 μS
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • traceable conductivity standard

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use appropriate media to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : potassium chloride,N-propanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers