Learn More
Fisherbrand™ Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Tool-free rotors and quick-spin options make the Fisherbrand Mini-Centrifuge fast and easy to use. The locking lid is functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction. 
Brand:  Fisherbrand™ HS120336
Product Code. 15358266
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The Fisherbrand Mini-Centrifuge is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 mL) for customized use. The centrifuge housing is made from ABS plastic and the lid is a polycarbonate plastic enabling the unit to be easily cleaned. A braking function quickly stops the unit when powered off or when the lid is opened.
  • A locking lid for added protection when the unit is in use
  • A larger lid to accommodate longer and skirted tubes, as well as taller tube lids
  • A braking function to quickly stop the unit when powered off or when the lid is opened
  • Meets the latest applicable North American & EU safety requirements
  • Weighted rubberized base eliminates potential movement and further dampens vibration and noise
Services
Five Years
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products