missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Silicone Bulb-Type Safety Pipet Fillers
Chemical-resistant, resilient, silicone pipet fillers with glass valves improve worker safety
£20.49
Specifications
Compatible Tips | No |
---|---|
Adjustable Tip Spacing | 4.5 mm to 9 mm |
Autoclavable | Not Autoclavable |
For Use With | Pipette |
Material | Silicone |
Description
Improve worker safety with these chemical-resistant, resilient, silicone pipet fillers with glass valves.
- Glass valves improve worker safety by eliminating mouth pipetting and handling of hazardous materials
- Long-lasting silicone is highly resistant to organic solvents
- Fit any pipet with O.D. ranging from 0.19 to 0.38 in. (4.5 to 9 mm)
To Use:
- Fill and dispense pipets with one hand
- Expel air by squeezing valve '1' above bulb
- Draw up liquid by pressing valve '2' on stem
- To release, press valve '3' on side of stem
Specifications
No | |
Not Autoclavable | |
Silicone | |
1 | |
50 mL |
4.5 mm to 9 mm | |
Pipette | |
3 Valve Standard | |
Pipette Filler | |
Pipet |