Learn More
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
POA
Specifications
Description | Top style: Heavy duty tooled with capacity scale |
---|---|
Material | Borosilicate Glass |
No. of Positions | 1 |
Writing Surface | Matte Finish Ceramic Enamel Marking |
Disposable | No |
Description
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
- Capacity scale measures approximate volume
- Matte-finish ceramic marking spot takes writing and erases easily
Compliance
- Designed from ASTM™ Specification E1404, Type I, Class 2 requirementsr
Specifications
Top style: Heavy duty tooled with capacity scale | |
1 | |
No |
Borosilicate Glass | |
Matte Finish Ceramic Enamel Marking | |
Wide |