missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heavy-Duty/Utility Funnels

Ideal for continuous lab work and heavy-duty jobs

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 1.40000kg

 View more versions of this product

Product Code. 11997884

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
  • Autoclavable to 120°C (248°F)

Specifications

Specifications

Yes
500mL
PP (Polypropylene)
20.5cm
20.5cm
Videos
Documentation