missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids

Fit standard spectrophotometer cell holders

Brand:  Fisherbrand™ 14958105

 View more versions of this product

Product Code. 15235993

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: PTFE lid or stopper
This item is not returnable. View return policy

Description

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Micro Fluorescence Cell
45 μL
PTFE
Ea., 2 Ea./Cs.
200 to 2500 nm
Standard spectrophotometer cell holder
Transmission from far ultraviolet through the infrared
Center H: 15mm
Quartz
3mm
3 mm
45 mm
12.5 mm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers