missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Borosilicate Glass Tubes with Polypropylene Screw Cap

Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 10421541

  • £555.00 / Pack of 1000


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Natural linerless polypropylene screw cap
  • With marking spot

Ideal for tissue culture, bacteriology, clinical chemistry, blood typing, and crossmatching procedures

Specifications

Specifications

16mm
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
Tray
16mL
125mm
Disposable Culture Tube
No
Yes
Marking Spot
250/Tray
Screw
Borosilicate Glass
Round Bottom
Videos
Documentation View accessory products